missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Small Molecule System Suitability Standard

Catalog No. PIA51740
Change view
Click to view available options
Quantity:
10 injections, 0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA51740 10 injections, 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA51740 Supplier Thermo Scientific™ Supplier No. A51740

The Thermo Scientific™ Pierce™ Small Molecule System Suitability Standard provides a pre-formulated mixture of nine small-molecule standards to assess system performance in both positive and negative ionization modes for TSQ Triple Quadrupole and Orbitrap Exploris™ mass spectrometers.

  • A pre-formulated mixture of nine small-molecule standards to assess system performance in both positive and negative ionization modes for TSQ Triple Quadrupole and Orbitrap Exploris mass spectrometers
  • Comprehensive mixture of nine small-molecule compounds (mass range: 76 to 1279)
  • Designed to assess instrument performance in both positive and negative ionization modes
  • Highly validated, high-quality, rigorously-tested formulation with lot-specific certificates of analysis
  • Convenient, pre-formulated ready-to-use liquid format
  • An excellent QC assay tool for quality assessment of LC and MS instrument status

Specifications

Color Colorless
Quantity 10 injections, 0.25 mL
Grade MS Grade
Physical Form Liquid
In which mode can I run the Small Molecule Suitability Standard?

The Small Molecule Suitability Standard can be run in Positive or Negative mode.

Does the Small Molecule Suitability Standard need to be diluted?

The Small Molecule Suitability Standard can be diluted 1:10 (25 µL of standard with 225 µL of 0.1% Formic Acid) or can be used without dilution. Glass vials are provided for dilutions. It is important to use 0.1% Formic Acid solution for dilution, in order to keep all components in solution.

What is the mass range for the Small Molecule Suitability Standard?

The mass range for the Small Molecule Suitability Standard (SMSS) is 76 to 1279 m/z.