missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Dimethylformamide (DMF), Sequencing grade

Catalog No. PI20673
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20673 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20673 Supplier Thermo Scientific™ Supplier No. 20673

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform liquid chromatography (HPLC) and mass spectrometry (LC-MS) applications with this 99.5%-pure DMF solvent (MW 73.09, CAS 68-12-2) with low UV-absorptivity.

Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.

This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps

Product Details:

  • General properties of DMF
  • Alternative names: n,n-Dimethylformamide, n-Formyldimethylamine
  • Molecular formula: C3H7ON
  • Molecular weight: 73.09
  • Density:0.945g/mL
  • CAS number: 68-12-2

Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

Specifications

Item Name Dimethylformamide (DMF)
Quantity 50 mL
Content And Storage Store at room temperature.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce
For Use With HPLC, Mass Spectrometry

For Research Use Only. Not for use in diagnostic procedures.