missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Rapid Antibody Isotyping Kit - Mouse

Catalog No. pi26178
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26178 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26178 Supplier Thermo Scientific™ Supplier No. 26178
Only null left

Identify mouse monoclonal antibody class and subclass in 5 minutes with these single-use, cassette-based lateral-flow assays.

Thermo Scientific™ Pierce Rapid Mouse Antibody Isotyping Kits are single-use, cassette-based kits for 5-minute lateral-flow assay determination of mouse monoclonal antibody class and subclass identity.

The kits are composed of small cartridges that provide a color-readout of the monoclonal antibody isotype within five minutes after pipetting a small amount of diluted culture supernatant or ascites fluid sample into the sample well. The kits determine mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM.

The assay is performed by simply adding a properly diluted tissue culture supernatant or mouse ascites sample to the well of the small cassette. Gold conjugates embedded in the cassette form specific class- and subclass-soluble complexes with the antibodies in the sample. These complexes travel the length of the membrane and are resolved on the anti-isotype and class-specific antibody-impregnated membrane. Results are displayed as a red band indicating the antibody isotype or subclass.

Highlights:

  • Long shelf life – stable for more than 1 year at room temperature

  • Single-step – add diluted antibody sample to the loading-well of cassette

  • Fast – within 5 minutes a band appears that indicates the antibody isotype

  • Sensitive – detect antibodies in tissue culture media or ascites fluid samples at any concentration greater than 10ng/mL

  • Specific – assay does not crossreact with fetal bovine serum (FBS)

  • Reliable – results are similar to the standard ELISA-based isotyping assays

Includes:

Ten pouches, each containing two (Part No. 26178) or three (Part No. 26179) cassettes, plus a bottle of sample diluent

Specifications

Description Pierce Rapid Isotyping Kit - Mouse
Product Line Pierce
Target Specificity Target-specific
Quantity 10 Tests
For Use With (Application) Membrane Blot
Assay Isotyping Assay
Detection Method Chromogenic
Content And Storage Sufficient For: 10 isotype determinations
• Isotyping Cassettes (two per pouch), 10 pouches
• Sample Diluent, 45 mL

Store kit at 2–8°C.

For Research Use Only. Not for use in diagnostic procedures.