Learn More
Thermo Scientific™ Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well
The endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.
Supplier:  Thermo Scientific™ A43888
Catalog No. PIA43888
Only null left
Explore available promotions
Description
Description
These endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.
Specifications
Specifications
Strip Plate
Sterile
5 plates, 1 frame
Endotoxin level <0.0005 EU/well. 12 strip x 8 well 96-well plate, strips are individual wrapped.
Chromogenic LAL Endotoxin Assay, Kinetic Chromogenic LAL Assay and Kinetic Turbidimetric LAL Assay
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.