Learn More
Thermo Scientific™ Pierce™ Pyrogen-Free Glass Test Tubes
Catalog No. PIA43889
Change view
Click to view available options
Quantity:
30 Tubes
Catalog No. Quantity
PIA43889 30 Tubes
1 options
Catalog No. PIA43889 Supplier Thermo Scientific™ Supplier No. A43889
Only null left
The endotoxin-free glass test tubes contain less than 0.005 EU/mL endotoxin. Recommended for use as reaction tubes in Gel Clot assays.
These endotoxin-free glass test tubes contain less than 0.005 EU/mL endotoxin. They are recommended for use as reaction tubes in endotoxin detection assays.
Specifications
Description Endotoxin level <0.005 EU/mL
Material Glass
For Use With Gel Clot and Chromogenic Endotoxin Assay
Quantity 30 Tubes
Product Type Test Tube
What are the dimension of the Pierce Pyrogen-Free Glass Test Tubes (Cat. No. A43889)?
The Pierce Pyrogen-Free Glass Test Tubes have an outer diameter of 13 mm and a length of 100 mm.
For Research Use Only. Not for use in diagnostic procedures.