missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein Coated Plates

Catalog No. PI15132
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Surface Treatment:
Protein A
Protein A/G
Protein G
Protein L
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Surface Treatment
PI15132 Clear Strip Plate Protein A
PI15130 Clear Standard Protein A
PI15155 Black Standard Protein A
PI15154 White Standard Protein A
PI15133 Clear Strip Plate Protein G
PI15131 Clear Standard Protein G
PI15157 Black Standard Protein G
PI15156 White Standard Protein G
PI15138 Clear Strip Plate Protein A/G
PI15190 Clear Standard Protein L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. PI15132 Supplier Thermo Scientific™ Supplier No. LSG15132

Thermo Scientific™ Pierce Protein A, G, A/G and L Coated Plates provide an alternative to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques.

These plates are uniformly and stably coated with one of four popular immunoglobilin-binding proteins (Protein A, Protein G, Protein A/G, or Protein L). They enable antibodies, or pre-established antibody-antigen complexes, to be captured to wells of the polystyrene plate for subsequent detection in plate readers. The strategy is useful for a variety of assay situations, but it is generally not suitable for sandwich-type ELISA.

Thermo Scientific Pierce Protein Coated Plates provide an alternative to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques.

Highlights
• Retain antibody activity, which can be lost when antibodies are immobilized by passive adsorption
• Orient antibodies for maximum antigen-binding capacity
• Immobilize antibodies for plate assays without prior purification
• Ensure minimal variation (<5% well-to-well) from consistent coating
• Reduce nonspecific binding because plates are pre-blocked with SuperBlock Blocking Buffer
• Bind 2 to 5 pmol of antibody per well (based on tests with rabbit IgG)

Protein A/G Coated Plates
• Fusion protein containing four antibody-binding sites from Protein A and two from Protein G
• Binds to most IgG species because it combines the specificities of Protein A and Protein G
• Binds to Fc region of antibodies, ensuring optimal orientation
• Binds well using a wide range of pH

Protein A Coated Plates
• Binds strongly to IgG from human, rabbit, guinea pig, pig, dog and rhesus monkey
• Binds strongly to mouse IgG2a, IgG2b and IgG3
• Binds to Fc region of antibodies for optimal orientation

Protein G Coated Plates
• Binds strongly to IgG from many species including human, mouse, rabbit, sheep, and goat
• Binds only to IgG—no cross-reactivity with other antibody classes
• Binds to Fc region of antibodies for optimal orientation

Protein L Coated Plates
• Binds to all classes of immunoglobulins (IgG, IgM, IgA, IgE and IgD)
• Binds to the VL region of kappa light chains (human I, III, IV and Mouse I) without interfering with antigen-binding sites
• Binds to Fab fragments and ScFv
• Does not bind bovine, goat or sheep immunoglobulin
• Binds weakly to rabbit immunoglobulin
• Pre-blocked to reduce nonspecific binding

Specifications

Binding Property Standard Capacity, Affinity
Format Strip Plate
Content And Storage Note: Plates are activated to 1 μL and supplied pre-blocked with Thermo Scientific SuperBlock Blocking Buffer. Use these plates for single-antibody assays only. In multiple antibody assays, such as sandwich ELISAs, the first antibody cannot block all binding sites and subsequent antibodies will bind to the plate, resulting in false positives.

Upon receipt store plates at 4°C in unopened pouches. Place opened, unused plates in a resealable bag and store desiccated at 4°C. Plates are shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment Protein A
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less