Learn More
Thermo Scientific™ SDAD (NHS-SS-Diazirine) (succinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate)
Description
Thermo Scientific™ Pierce NHS-Diazirine Crosslinkers combine proven NHS-ester and diazirine-based photoreaction chemistries to conjugate amine-containing molecules with nearly any other functional group via long-wave UV-light activation.
Highlights:
Versatile - different amine-reactive diazirine derivatives capture specific intracellular or cell-surface protein interactions
Controllable - two-step chemical crosslinking is activated using common laboratory UV lamps
Reversible - disulfide-containing spacers enable cleavage using reducing agents
Easy - reagents are photo-stable under typical laboratory lighting conditions so there is no need to perform experiments in the dark
Better than aryl azides - diazirine photo-reactive group has better photostability and is more efficiently activated by long-wave UV light
Specifications
Specifications
| Chemical Reactivity | Amine-Nonselective |
| Cleavable | By Thiols |
| Description | SDAD |
| Molecular Weight (g/mol) | 388.46 |
| PEGylated | No |
| Spacer Arm Length | 13.5 Å |
| Content And Storage | Upon receipt store product at 4°C protected from light and moisture. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.