missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NeutrAvidin™ Protein, Maleimide-Activated

Catalog No. pi31007
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31007 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31007 Supplier Thermo Scientific™ Supplier No. 31007
Only null left

Purified NeutrAvidin protein for Western blot, ELISA or other biotin-based detection methods.

Thermo Scientific™ NeutrAvidin Protein is a specially-prepared form of avidin biotin-binding protein that decreases background in Western blotting and ELISA; available items include standard HRP, AP and fluorescent conjugates for biotin-based assays.

NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Purified and conjugated forms of NeutrAvidin Protein provide exceptional performance in Western blot, ELISA and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with NeutrAvidin Protein are generally equivalent or better than with the significantly more expensive streptavidin.

Highlights:

  • Near-neutral isoelectric point – pI equals 6.3, more neutral than native avidin

  • Nearly devoid of glycosylation – decreased possibility of lectin binding compared to native avidin

  • No RYD recognition sequence – no known off-target binding domains like streptavidin

  • Affordable – significantly less expensive than streptavidin

  • Available in multiple formats – choose purified NeutrAvidin Protein, prepared HRP and AP conjugates or activated forms for a variety of techniques

Specifications

Conjugate Streptavidin
Quantity 5 mg
Content And Storage Upon receipt store product 4°C. Product is shipped at ambient temperature.
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Solubility Soluble in Water
Form Powder
Product Line NeutrAvidin
Product Type NeutraAvidin (Reactive)
Label or Dye Streptavidin

For Research Use Only. Not for use in diagnostic procedures.