Learn More
Thermo Scientific™ Pierce™ Midi Gel Power Staining Kit
The Thermo Scientific Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Midi Gel Power Staining Kit enables rapid Coomassie dye staining of proteins in midi-size polyacrylamide gels and the removal of unbound stain from the gel matrix.
Supplier: Thermo Scientific™ LSG22839
Description
The Thermo Scientific Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Midi Gel Power Staining Kit enables rapid Coomassie dye staining of proteins in midi-size polyacrylamide gels and the removal of unbound stain from the gel matrix.
- High performance—performance is equivalent to, or better than, traditional Coomassie dye staining techniques
- Fast—optimized ionic solutions enable rapid Coomassie dye staining of protein and gel destaining.
- Compatible—verified to work with commonly used precast and homemade SDS-PAGE gels
Traditional Coomassie dye staining techniques require staining for one hour to overnight and prolonged destaining for desired results. When used together, the Pierce Power stain solution, destain solution, and gel pads enable efficient staining and destaining in minutes with results equivalent to, or better than, traditional Coomassie dye staining techniques.
Specifications
| Note: The kit contains sufficient reagents to stain 15 midi-sized gels at 30 mL each. Contents: Midi Staining Gel Pads, 2 boxes of 15 each (8 sheets/pad) Pierce Power Stain Solution, 480 mL Destain Solution, 480 mL |
|
| In-Gel Detection | |
| Coomassie | |
| Proprietary gel stain and destain solutions | |
| 15 gels | |
| Protein |
| Midi Gel Power Staining Kit, 15 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 15 Midi-Sized Gels at 30mL Each | |
| Colorimetric | |
| Fast Protein Gel Stain | |
| Pierce | |
| 15 Midi-sized gels at 30mL each |
For Research Use Only. Not for use in diagnostic procedures.