Learn More
Thermo Scientific™ Pierce™ Maleimide Activated Plates
Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
Supplier: Thermo Scientific™ 15152
Description
Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights• Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
• Pre-blocked to reduce nonspecific binding
Chemical Identifiers
| Standard Capacity, Covalent | |
| Pierce | |
| 100 μL | |
| Maleimide | |
| Blocker™ BSA Buffer |
| Note: Plates are supplied pre-blocked with bovine serum albumin. Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature. |
|
| Polystyrene | |
| 96 | |
| 5 Plates | |
| 5 plates |
Specifications
| Standard Capacity, Covalent | |
| Note: Plates are supplied pre-blocked with bovine serum albumin. Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature. |
|
| Pierce | |
| Detection Plate, Immunoassay, ELISA | |
| 100 μL | |
| Maleimide | |
| Blocker™ BSA Buffer |
| Standard | |
| Chemiluminescence | |
| White | |
| Polystyrene | |
| 96 | |
| 5 Plates | |
| 5 plates |