missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Maleimide Activated Plates

Catalog No. PI15150
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Detection Method
PI15150 Clear Strip Plate Colorimetric
PI15152 White Standard Chemiluminescence
PI15153 Black Standard Fluorescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI15150 Supplier Thermo Scientific™ Supplier No. 15150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.

Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights
• Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
• Pre-blocked to reduce nonspecific binding

Specifications

Binding Property Standard Capacity, Covalent
Format Strip Plate
Content And Storage Note: Plates are supplied pre-blocked with bovine serum albumin.

Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 96
Surface Treatment Maleimide
No. per Pack 5 Plates
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less