missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CHCA MALDI Matrix, Single-Use

Catalog No. pi90031
Change view
Click to view available options
Quantity:
24 x 1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI90031 24 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI90031 Supplier Thermo Scientific™ Supplier No. LSG90031
Only null left

Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).

Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.

The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.

Highlights:

  • Highly purified, recrystallized reagent
  • Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
  • Provides accurate quantity of matrix every time

Specifications

Description Pierce CHCA MALDI Matrix
For Use With (Equipment) Mass Spectrometer
Product Type MALDI Matrix
Content And Storage Upon receipt store product at 4°C.
Starting Material Small Molecules
Final Product Type Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 24 x 1 mg
Form Solid

For Research Use Only. Not for use in diagnostic procedures.