missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glycoprotein Staining Kit

Catalog No. PI24562
Change view
Click to view available options
Quantity:
750 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24562 750 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI24562 Supplier Thermo Scientific™ Supplier No. 24562

missing translation for 'validMainframeMsg'

Rapidly stain glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method in this convenient kit.

Thermo Scientific™ Pierce Glycoprotein Staining Kit is a convenient, fast and sensitive kit for staining glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method.

The kit provides three essential reagents and a complete protocol to specifically stain glycosylated proteins (glycoproteins) that have been separated by polyacrylamide gel electrophoresis (PAGE). A gel or membrane containing separated proteins is treated with a periodate solution, which oxidizes cis-diol sugar groups in glycoproteins. The resulting aldehyde groups are detected through the formation of Schiff-base bonds with a reagent that produces magenta bands. Traditionally called the periodic acid-Schiff (PAS) reagent method, this staining technique is made easier by the convenient packaging of essential reagents in one kit with easy-to-follow instructions. The kit includes positive and negative control proteins.

Highlights:

  • Specific for glycoproteins – periodic acid-Schiff (PAS) method specifically detects glycosylated proteins having sialic acid and other oxidizable carbohydrate groups

  • Optimized protocol – the traditional three-reagent method has been streamlined to achieve optimal staining in less than two hours (half the time required for other staining methods)

  • Colorimetric – glycoproteins are detected as magenta bands against a faint pink or colorless gel background

  • Qualitative staining – detection limit depends on the extent of glycosylation and protein size and abundance; detects 0.625ng avidin and 0.16g horseradish peroxidase per band

  • Controls included – complete kit includes positive and negative control proteins (HRP and STI, respectively).

  • Gels and membranes – effective for use with polyacrylamide mini gels or nitrocellulose membranes

Includes:

Three reagents in 250mL bottles (1 solution and 2 dry powders to which water must be added), plus two vials of lyophilized proteins (1mg each)

Specifications

Content And Storage Sufficient For: 10 mini gels (SDS-PAGE)
• Oxidation Reagent, 2.5 g powder to make 250 mL
• Reduction Reagent, 1.25 g powder to make 250 mL
• Positive Control (Horseradish Peroxidase), 1 mg
• Negative Control (Soybean Trypsin Inhibitor), 1 mg
• Glycoprotein Stain, 250 mL

Store all components at 4°C.
Detection Location In-Blot Detection, In-Gel Detection
Detection Method Colorimetric
Label or Dye Fuchsin Sulfite
Quantity 750 mL
Target Molecule Proteins (Glycoproteins)
Product Line Pierce
Product Type Glycoprotein Staining Kit

For Research Use Only. Not for use in diagnostic procedures.