missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glutathione Coated Plates

Catalog No. p-7227136
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format Detection Method
PI15340 Black Standard Fluorescence
PI15140 Clear Strip Plate Colorimetric
PI15240 White Standard Chemiluminescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI15340 Supplier Thermo Scientific™ Supplier No. LSG15340
Only null left

Thermo Scientific™ Pierce Glutathione Coated Plates are standard 96-well microplates coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins.

Pierce Glutathione Coated Plates are coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins. These coated plates can be used in the analysis of cell lysates to determine the presence and concentration of the GST-fusion protein produced in an expression system. Plates can also be used to capture and coat fusion proteins via their GST tag for subsequent binding assays involving the recombinant protein. As the plates are pre-blocked, initial purification of the cell lysates is not necessary.

Thermo Scientific Pierce Glutathione Coated Plates are microplates coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins.

Features of Glutathione Coated Plates include:
Selective—specifically binds fusion proteins containing glutathione S-transferase
Capacity—binds approx. 10 ng GST protein per well
Versatile—ready-to-bind plate format is useful for protein interaction studies
Compatible—detergents used to lyse cells do not inhibit GST protein binding to pre-coated plates (by contrast with direct coating methods with polystyrene plates)
Ready to use—Pre-blocked with SuperBlock Blocking Buffer to reduce nonspecific binding

Specifications

Binding Property Standard Capacity, Affinity
Format Standard
Content And Storage Store at 4°C in unopened pouches. Once opened, store unused plates in a resealable bag with desiccant at 4°C.
Detection Method Fluorescence
Packaging 5 Plates
Product Line Pierce
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Glutathione
Plate Blocking SuperBlock™ Buffer
Show More Show Less