missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fast Western Blot Kit, ECL Substrate

Catalog No. PI35055
Change view
Click to view available options
Quantity:
1 L Kit
200 mL Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI35055 200 mL Kit
PI35050 1 L Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI35055 Supplier Thermo Scientific™ Supplier No. 35055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized reagents that shorten the time to perform a typical Western blot from 4 hours to approximately 55 minutes.

Thermo Scientific™ Pierce Fast Western Blot Kit, ECL Substrate contains optimized reagents that shorten the time to perform a typical Western blot from 4 hours down to approximately 55 minutes.

The kit provides all the reagents necessary to complete a Western blot being probed with a mouse or rabbit primary antibody. The protocol requires minimal hands-on time and yields results comparable to classic Western blotting with ECL. The included Pierce ECL Substrate produces a chemiluminescent signal, which is detected using photographic or other imaging methods. Blots can be repeatedly exposed to film to obtain optimal results or stripped of the primary antibody and immunodetection reagents and reprobed.

Highlights:

  • Fast – all the sensitivity of Pierce ECL substrate; saves three to four hours over traditional blotting methods

  • Convenient – no expensive hardware or extra bench space needed; eliminates membrane clogging issues

  • Economical – costs about the same as a classical Western blot; no expensive consumables or extra equipment

  • Easy – optimized kits includes all reagents except primary antibody

  • Versatile – kits work with both rabbit and mouse primary antibodies

Includes:

Kits contain all reagents to perform Western blots probed with mouse and rabbit primary antibodies, including: Fast Western Antibody Diluent; Fast Western 10X Wash Buffer; Fast Western Optimized HRP Reagent; Pierce ECL Detection Reagent 1; Pierce ECL Detection Reagent 2

Specifications

Product Type Western Blot Kit
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce
Quantity 200 mL Kit
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Mouse, Rabbit
Sufficient For 5 Mini Blots
Content And Storage Sufficient For: 5 mini blots probed with mouse or rabbit primary antibodies
• Fast Western Antibody Diluent, 100 mL
• Fast Western 10X Wash Buffer, 50 mL
• Fast Western Optimized HRP Reagent, 5 mL
• Pierce ECL Detection Reagent 1, 25 mL
• Pierce ECL Detection Reagent 2, 25 mL

Store at 4°C.
Show More Show Less
Does my primary antibody need to be conjugated for using with the Pierce Fast Western Blot Kit, ECL Substrate (Cat. No. 35050, 35055)?

No, your primary antibody does not need to be conjugated for using with the Pierce Fast Western Blot Kit, ECL Substrate (cat. No. 35050, 35055). The kit includes all necessary reagents except the primary antibody, and it works with both mouse and rabbit primary antibodies. The HRP reagent directly binds to both mouse and rabbit primary antibodies, thus, a secondary antibody is not needed.

For western blotting with the Pierce Fast Western Blot Kit, ECL Substrate (Cat. No. 35050), do I need a secondary antibody?

The kit contains the horseradish peroxidase (HRP) reagent and ECL detection reagent, therefore the kit works with blots probed with mouse or rabbit primary antibodies. The HRP reagent directly binds to both mouse or rabbit primary antibodies, thus, a secondary antibody is not needed.

For Research Use Only. Not for use in diagnostic procedures.