Learn More
Thermo Scientific™ Ethylene Glycol
Catalog No. PI29810
Change view
Click to view available options
Quantity:
200 mL
Catalog No. Quantity
PI29810 200 mL
1 options
Catalog No. PI29810 Supplier Thermo Scientific™ Supplier No. 29810
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specially purified 50% ethylene glycol solution to provide antifreeze protection and freezer storage stability for antibody-enzyme conjugates.
Thermo Scientific Pierce Ethylene Glycol Solution provides exceptional antifreeze protection and storage stability for antibody-enzyme conjugates because it is purified to remove impurities commonly found traditional glycerol stocks.
Highlights:
  • Specially purified to remove impurities such as aldehydes, peroxides, iron and UV absorbing hydrocarbons
  • Suitable for enzyme storage without the worry of losing enzymatic activity
  • Stable for months
Specifications
Quantity 200 mL
Physical Form Liquid
For Research Use Only. Not for use in diagnostic procedures.