missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ethidium Bromide Solution (10 mg/mL)

Catalog No. PI17898
Change view
Click to view available options
Quantity:
5 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17898 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17898 Supplier Thermo Scientific™ Supplier No. LSG17898
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visualize DNA fluorescently in a variety of molecular biology applications using these dilute (0.625mg/mL) or concentrated (10mg/mL) ethidium bromide solutions.

Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.

Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.

Highlights:

0.625mg/mL EtBr Solution:

  • Each drop contains 25μg of ethidium bromide and is ideal for a 50mL agarose gel
  • After adding one drop of solution per 50mL of gel, the final concentration of EtBr will be 0.5μg/mL, which is the recommended concentration for electrophoresis of nucleic acids
  • Can also be used as the running buffer or for staining gels after electrophoresis

10mg/mL EtBr Solution:

  • A safe and convenient way to use ethidium bromide while minimizing exposure

Specifications

Content And Storage Store at room temperature, protected from light.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Ethidium Bromide
Quantity 5 x 10 mL
Target Molecule RNA, DNA
Product Type Ethidium Bromide Solution
Do you have a protocol for ethidium bromide staining in agarose gels?

For use in agarose gels:

- Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. Do not melt agarose that already contains ethidium bromide.

For staining agarose gels after electrophoresis:

- You can stain gels that have been run in the absence of ethidium bromide by covering the gel in 0.5 µg/mL ethidium bromide in water and gently agitating for 10 to 30 minutes.
- If necessary, gels can be destained by shaking in H2O for an additional 30 minutes. To stain RNA gels, you should minimize staining time and definitely include a destaining period.

What can ethidium bromide be used to detect and what is its sensitivity?

Ethidium bromide can be used to detect ssDNA, RNA, and dsDNA. This fluorescent dye intercalates between the stacked bases of nucleic acids, and exhibits an increased fluorescence at 590 nm. Ethidium bromide can detect down to ~1-10 ng/band of dsDNA in an agarose gel.

For Research Use Only. Not for use in diagnostic procedures.