missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Eau sans endotoxines
Description
Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including proteases.
Using Endotoxin-Free Water helps eliminate the risk of harmful activity produced by bacteria that can interfere with cell health and function. It is suitable for usage in molecular biology or general biochemistry applications and contains <0.005 EU/mL endotoxin.
Spécifications
Spécifications
| Aperçu général | Eau sans endotoxines |
| Endotoxine | <0,005 EU/mL |
| Emballage | Bouteille(s) |
| Stérilité | Stérile |
| Quantité | 4 x 50 mL |
| Méthode de purification | Filtré stérile avec un filtre de 0,5 μm |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.