missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Endotoxin-Free Magnesium Chloride (MgCl2) (10 mM)

Catalog No. PIA43884
Change view
Click to view available options
Quantity:
5 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA43884 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA43884 Supplier Thermo Scientific™ Supplier No. A43884
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Pierce Endotoxin-Free MgCl2 buffer is used to prepare samples for any LAL assay. As a diluent, it will eliminate the inhibitory chelation effects of EDTA. Contains <0.005 EU/mL endotoxin.

Pierce Endotoxin-Free MgCl2 is suitable for the preparation of samples for endotoxin detection (LAL) assay. As a diluent, it will eliminate the inhibitory chelation effects of EDTA. It contains <0.005 EU/mL endotoxin.

Specifications

Description Endotoxin-Free MgCl2 buffer
Concentration 10 mM
For Use With (Equipment) LAL Endotoxin Assay
Quantity 5 x 10 mL
Packaging Vial

For Research Use Only. Not for use in diagnostic procedures.