Learn More
Thermo Scientific™ Pierce™ Dilution-Free™ Tubestrip Holder
Catalog No. fera57184
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
FERA57184 1 each
1 options
Catalog No. FERA57184 Supplier Thermo Scientific™ Supplier No. A57184
Only null left
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. Protein standard concentrations are printed on the Tubestrip Holder for easy reference and pipetting on the benchtop.
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. Protein standard concentrations are printed on the Tubestrip Holder for easy reference and pipetting on the benchtop.
Pierce Dilution-Free BSA Protein Standards
Pierce Dilution-Free BSA Protein Standards are high-quality bovine serum albumin (BSA) reference samples for generating accurate standard curves and calibration controls in total protein assays. They are packaged as a set of seven pre-diluted standards in a multichannel pipette-friendly tubestrip, eliminating the need for serial dilutions. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.
Specifications
Product Line Pierce Dilution-Free
Product Type Tubestrip holder
Quantity 1 each
For Research Use Only.