missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Coomassie Brilliant Blue Dyes

Catalog No. p-7229246
Change view
Click to view available options
Label or Dye:
Coomassie Brilliant Blue G-250
Coomassie Brilliant Blue R-250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
PI20278 Coomassie Brilliant Blue R-250
PI20279 Coomassie Brilliant Blue G-250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20278 Supplier Thermo Scientific™ Supplier No. LSG20278
Only null left

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains.

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains. Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Typically, coomassie gel stains and protein assay reagents are formulated as very acidic solutions in 25 to 50% methanol. In acidic conditions, the dye binds to proteins primarily through basic amino acids (primarily arginine, lysine, and histidine), and the number of coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Protein binding causes the dye to change from reddish-brown to bright blue (absorption maximum equals 595 nm).

Features include:

  • Easy detection—develops intensely colored complexes with proteins
  • High sensitivity—can determine as little as 0.5 μg/cm2 of protein present in a gel matrix
  • Reversible staining—anion of Coomassie Brilliant Blue dye formed in the acidic staining medium combines with the protonated amino groups of proteins by electrostatic interaction; resulting complex is reversible under proper conditions
  • Differentiation between bound and unbound dye—when dissolved in 0.01 M citrate buffer at pH 3.0, has an absorption maximum at 555 nm; protein-dye complex is characterized by a peak slightly broader than that of free dye with a maximum at 549 nm

Specifications

Content And Storage Store at room temperature.
Detection Location In-Gel Detection, In-Solution Detection, In-Blot Detection
Detection Method Colorimetric
Label or Dye Coomassie Brilliant Blue R-250
Quantity 50 g
Target Molecule Protein
Product Line Pierce
Product Type Protein Gel Stain