missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Coomassie Brilliant Blue Dyes

Catalog No. PI20279
Change view
Click to view available options
Label or Dye:
Coomassie Brilliant Blue G-250
Coomassie Brilliant Blue R-250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
PI20279 Coomassie Brilliant Blue G-250
PI20278 Coomassie Brilliant Blue R-250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20279 Supplier Thermo Scientific™ Supplier No. LSG20279

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains.

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains. Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Typically, coomassie gel stains and protein assay reagents are formulated as very acidic solutions in 25 to 50% methanol. In acidic conditions, the dye binds to proteins primarily through basic amino acids (primarily arginine, lysine, and histidine), and the number of coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Protein binding causes the dye to change from reddish-brown to bright blue (absorption maximum equals 595 nm).

Features include:

  • Easy detection—develops intensely colored complexes with proteins
  • High sensitivity—can determine as little as 0.5 μg/cm2 of protein present in a gel matrix
  • Reversible staining—anion of Coomassie Brilliant Blue dye formed in the acidic staining medium combines with the protonated amino groups of proteins by electrostatic interaction; resulting complex is reversible under proper conditions
  • Differentiation between bound and unbound dye—when dissolved in 0.01 M citrate buffer at pH 3.0, has an absorption maximum at 555 nm; protein-dye complex is characterized by a peak slightly broader than that of free dye with a maximum at 549 nm

Specifications

Content And Storage Store at room temperature.
Detection Location In-Gel Detection, In-Solution Detection, In-Blot Detection
Detection Method Colorimetric
Label or Dye Coomassie Brilliant Blue G-250
Quantity 50 g
Target Molecule Protein
Product Line Pierce
Product Type Protein Gel Stain
What is the absorption maximum of free and protein-bound dye Coomassie R-250?

When dissolved in 0.01 M citrate buffer at pH 3.0, it has an absorption maximum at 555 nm; the protein-dye complex is characterized by a peak slightly broader than that of the free dye with an absorption maximum at 549 nm.

To what amino acids does Coomassie R-250 bind?

In acidic conditions, Coomassie dye primarily binds basic amino acids (arginine, lysine and histidine).

How do I prepare a solution of Coomassie R-250 dye?

Add 100 mL of glacial acetic acid to 450 mL distilled water.
Dissolve 3 g of Coomassie R-250 dye in 450 mL methanol.
Mix the acetic acid and methanol solutions.
Filter the solution before use.

Is Coomassie Brillant Blue R-250 available already prepared in solution?

Yes, Imperial Protein Stain (Cat. Nos. 24615 (1L) and 24617 (3 x 1L) is a ready-to-use colorimetric stain formulated with Coomassie R-250 dye that delivers consistent nanogram-level detection of proteins in polyacrylamide electrophoresis gels.

What are the differences between R-250 and G-250 Coomassie Dyes?

Coomassie G-250 has an additional methyl group on the lower two central rings in the structure, as compared to Coomassie R-250. Bands stained with Coomassie R-250 are reddish-purple in color; Coomassie G250 bands are greenish-blue in color.

Sometimes referred to as colloidal Coomassie Blue, the G-250 form differs from R-250 in more readily forming dye-dye aggregates in solution. Consequently, G-250 interacts more weakly with polyacrylamide gel matrix than with itself, resulting in lower background that can be more easily washed away.

Can membranes be stained with the Colloidal Blue (G-250) Staining Kit after blotting?

No we do not recommend staining membranes with the Colloidal Blue (G-250) Staining Kit as the background will be too high. Better alternatives include:

- Invitrogen Reversible Membrane Protein Stain (Cat. No. IB7710): Allows for complete, reversible staining of protein on nitrocellulose & PVDF membranes. Sensitivity is higher than Ponceau S (<10 ng of BSA in 10 mins as blue bands) and the staining is reversible in 5 minutes. Western blot detection is unaffected by the staining and erasing process, and in some cases higher sensitivity is achieved.
- Coomassie (non-colloidal) staining: Stain in 0.1% Coomassie Blue R-250 in 50% methanol for 5 minutes and destain with several changes of 50% methanol and 10% acetic acid. Rinse with several changes of water, air dry and store for up to 12 months at −20 degrees C. Sensitivity is approximately at the 50-100 ng level.
- SimplyBlue SafeStain. There is a protocol included in the SimplyBlue SafeStain manual for staining dry PVDF membranes but it is not recommended for wet PVDF membranes or nitrocellulose because of the high background.
- Amido black: same as Coomassie stain but less sensitive. Please see protocol in the manual (https://tools.thermofisher.com/content/sfs/manuals/invitrolonpvdf_man.pdf).
- Ponceau S: same as Coomassie stain but less sensitive. Please see protocol in the manual (https://tools.thermofisher.com/content/sfs/manuals/invitrolonpvdf_man.pdf).
- UV transillumination: After blotting, place the membrane on a filter paper and allow to air-dry at room temperature for about 10 minutes. Rewet in 20% methanol and view the blot in front of white light while it is still wet; the bands will look more translucent than the membrane. If the bands disappear because the membrane is dry, rewet the membrane.