Learn More
Thermo Scientific™ Compat-Able™ Protein Assay Preparation Reagent Kit
Description
Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
Highlights:
Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
Simple and fast – prepare samples in less than 10 minutes with four simple steps
Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures
Includes:
Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay
Specifications
Specifications
| Content And Storage | Kit contains sufficient reagents to pretreat up to 500 samples • 250 mL Compat-Able Protein Assay Preparation Reagent 1 • 250 mL Compat-Able Protein Assay Preparation Reagent 2 Store at room temperature |
| Description | Compat-Able Protein Assay Preparation Reagent Set |
| Assay | BCA or Bradford Assay |
| Detection Method | Colorimetric |
| Product Line | Compat-Able |
| Product Type | Protein Quantitation Assay |
| Specificity | Not Target-Specific |
| For Use With (Application) | Incompatibility Elimination |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Quantity | 500 mL |
For Research Use Only. Not for use in diagnostic procedures.