missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Color Silver Stain Kit

Catalog No. pi24597
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24597 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI24597 Supplier Thermo Scientific™ Supplier No. LSG24597
Only null left

Detect proteins with this metallic silver staining technique that produces color variations to differentiate protein bands and spots in 1D and 2D gels.

Thermo Scientific™ Pierce Color Silver Stain Kit enables sensitive detection of proteins electrophoresed in 1D and 2D polyacrylamide gels using a metallic silver technique which produces color variations that facilitate differentiation of protein bands and spots.

The Color Silver Stain Kit contains four reagents and a flexible protocol for staining proteins in 1D and 2D electrophoresis gels. The method requires more time than monochromatic silver stains but provides greater flexibility in adapting the concentration of working solutions to obtain effective staining with different gel types and thicknesses and unusual protein targets. Proteins stain as black, blue, brown, red or yellow bands or spots according to a combination of their specific charge and other characteristics. Once results are known for a particular protein of interest, these slight color differences provide for easy band and spot identification.

Highlights:

  • Customizable – kit is a reagent set that allows customized formulation of working reagents and staining protocols for particular gel formats and silver-staining applications

  • Sensitive – detects many proteins at 0.1ng per square millimeter in standard-thickness gels

  • Broad protein detectability – stains a wider range of protein types than monochromatic silver stains (proteins that do not bind silver are revealed as yellow bands or spots)

  • Color differentiation – gel-separated proteins and polypeptide-containing macromolecules stain in five basic colors: black, blue, brown, red and yellow
  • Profiling resolution – multicolored detection of DNA, lipids and polysaccharides in addition to typical proteins provides additional landmarks to aid in 2D gel profiling

Includes:

Four reagent solutions (500mL each): silver, reducer base, reducer aldehyde and stabilizer base

Specifications

Content And Storage Kit contains sufficient reagents to stain 25 (18 cm2) or 40 (130 cm2) gels
• 500 mL Silver Reagent
• 500 mL Reducer Aldehyde Reagent
• 500 mL Reducer Base Reagent
• 500 mL Stabilizer Base Reagent
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye Silver
Quantity 2 L
Target Molecule Protein
Product Line Pierce
Product Type Stain Kit

For Research Use Only. Not for use in diagnostic procedures.