missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Background Eliminator Kit

Catalog No. PI21065
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21065 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21065 Supplier Thermo Scientific™ Supplier No. LSG21065
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly remove haze and artifacts from X-ray film to correct overexposed blots, whether from chemiluminescent Western blot or DNA or RNA autoradiography.

Thermo Scientific™ Pierce Background Eliminator provides fast, easy removal of haze and artifacts on developed X-ray film to correct for overexposure of blots, whether from chemiluminescent Western blotting or DNA or RNA autoradiography.

This two-reagent kit is simple to use. Mix the two reagents (1:1 ratio) and immerse a developed piece of film (recent or old) in the working solution. Watch for 1 to 5 minutes until the desired image appears, then stop the reaction by rinsing the film in water. Pierce Background Eliminator for Film evenly reverses development-signal over the entire film, preserving relative densitometry values so that analysis and publication of results is still valid and possible.

Highlights:

  • For overexposed X-ray film – reverses development of silver-based autoradiography film removing silver emulsion from the film surface

  • Recovers and preserved data – evenly reduces developed signal over the entire film so relative intensity (densitometry) of bands is not altered

  • Optimizes for signal-to-noise – allows readjustment of overdeveloped film to obtain the best image with respect to background haze, speckles and oversaturated bands that need to be resolved

  • Saves time and resources – eliminates the need to repeat experiments or re-expose a new piece of film to obtain an optimal image

Recommended for:

Remove background from any application that uses X-ray film exposures including: Western, Northern and Southern blots that use Thermo Scientific SuperSignal Substrates and the Pierce Chemiluminescent Hybridization and Detection Kit; In-gel detection systems; Gel-shift assays; Ribonuclease protection assays (RPA)

Specifications

Product Type Background Eliminator Kit
Quantity 200 mL
Detection Method Western Blot
Reactivity All species
Content And Storage Sufficient For: 2.5L of 1X solution to treat at least 10 films (13 x 18 cm ea.)
• Reagent A, 100 mL
• Reagent B, 100 mL

Store at room temperature
Product Line Pierce
Can I "fix" my overexposed film when using SuperSignal West Femto Maximum Sensitivity Substrate?

We offer the Pierce Background Eliminator Kit (Cat. No. 21065) that reduces high background, shading, speckling, and over-exposed bands on exposed film.

Can I "fix" my overexposed film when using SuperSignal West Dura Extended Duration Substrate?

We offer the Pierce Background Eliminator Kit (Cat. No. 21065) that reduces high background, shading, speckling, and over-exposed bands on exposed film.

For Research Use Only. Not for use in diagnostic procedures.