missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 8-Well Polystyrene Strip Plates, Corner Notch

Catalog No. PI15031
Change view
Click to view available options
Quantity:
100 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15031 100 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15031 Supplier Thermo Scientific™ Supplier No. 15031

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pierce 8-Well Strip Plates are optically clear, polystyrene plates that have a standard 96-well microplate footprint but also allow individual 8-well strips (i.e., plate columns) to be removed from the rigid frame.

Additional Information and Online Ordering

The format provides convenience and quality-assurance when performing partial-plate assays because unused wells can be removed to prevent confusion and cross-contamination.

Highlights:

  • Optically clear polystyrene strip-well construction for compatibility with standard plate readers
  • High binding capacity surface improves the overall performance in immunoassay applications
  • Certified for performance, having consistently low coefficients of variation (CV) among wells
  • Eight-well strips fit securely in the plate frame, remaining in place even if turned upside down
  • Guides help to avoid misorientation of the strip when returned to the holder
  • Strips are gently removed from the frame with a simple twist or a strip-well ejector

One Strip Well Ejector per package.

 

 

 

Specifications

Binding Property Adsorption, Hydrophilic
Content And Storage Contents: 100 plates, 5 plates per sealed tray
Detection Method Colorimetric
Packaging 100 Plate
Product Line Pierce
Color Clear
Product Type Well Plate
Material Polystyrene
No. of Wells 96
Surface Treatment Polystyrene
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 100 plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.