missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade

Catalog No. PI85172
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85172 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI85172 Supplier Thermo Scientific™ Supplier No. T85172K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

Thermo Scientific™ 0.1% Trifluoroacetic Acid in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

This 0.1% Trifluoroacetic Acid in Water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Validated

    – tested more than 20 ways to ensure maximal LC/MS sensitivity

  • Purified

    – processed, filtered and sealed to extend LC/MS column life

  • Convenient

    – packaged to eliminate variability and reduce handling

Specifications

Grade LC-MS
Item Name Trifluoroacetic Acid (TFA)
Quantity 1 L
Content And Storage Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Form Liquid
Reagent Type Solvent Mixture, Ion Pairing Reagent
Product Line Pierce
For Use With HPLC, Mass Spectrometry

For Research Use Only. Not for use in diagnostic procedures.