missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile, LC-MS Grade
Description
Thermo Scientific™ 0.1% Trifluoroacetic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
This 0.1% Trifluoroacetic Acid in Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.
Highlights:
Validated
– tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified
– processed, filtered and sealed to extend LC/MS column life
Convenient
– packaged to eliminate variability and reduce handling
Specifications
Specifications
| Grade | LC-MS |
| Item Name | Trifluoroacetic Acid (TFA) |
| Quantity | 1 L |
| Content And Storage | Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. |
| Form | Liquid |
| Reagent Type | Solvent Mixture, Ion Pairing Reagent |
| Product Line | Pierce |
| For Use With | HPLC, Mass Spectrometry |
For Research Use Only. Not for use in diagnostic procedures.