Learn More
Chemglass Life Sciences PIE-BLOCK System, Supplied with 1ea Safety Holder and 1ea of the Six (6) Listed Pie Wedges DFS Item
Supplier:  Chemglass Inc CG1991P01
PIE-BLOCK System, Supplied with 1ea Safety Holder and 1ea of the Six (6) Listed Pie Wedges. Aluminum construction provides excellent heat transfer while not interfering with the magnetic fieldHoles are spaced for optimum stirring of all vesselsHolder fits all 135mm diameter hotplate stirrersInserts are color coded for quick & easy identificationThermowell is provided for insertion of a contact thermometer or temperature sensorReaction PIE-BLOCK is designed to fit directly on 135mm diameter circular top hot plate stirrers. Versatile design of the safety pie plate holder allows up to four pie wedges on one hot plate & securely locks them together. All wedges are anodized aluminum & color-coded for quick & easy identification. A drilled thermowell is provided for insertion of contact thermometers or temperature sensors.Complete PIE-BLOCK System (CG-1991-P-01) Consists of 1 Each: CG-1991-P-05, CG-1991-P-10, CG-1991-P-11, CG-1991-P-12, CG-1991-P-13, CG-1991-P-14 and CG-1991-P-15
Catalog No. CG1991P01
Only null left
Explore available promotions