missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picolinamide, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 1452-77-3 |
|---|---|
| Molecular Formula | C6H6N2O |
| Molecular Weight (g/mol) | 122.13 |
| MDL Number | MFCD00023483 |
| InChI Key | IBBMAWULFFBRKK-UHFFFAOYSA-N |
| Synonym | picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59 |
| PubChem CID | 15070 |
| ChEBI | CHEBI:8200 |
| IUPAC Name | pyridine-2-carboxamide |
| SMILES | NC(=O)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC443360100
|
Thermo Scientific Chemicals
443360100 |
10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC443360500
|
Thermo Scientific Chemicals
443360500 |
50g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1452-77-3 | |
| 122.13 | |
| IBBMAWULFFBRKK-UHFFFAOYSA-N | |
| 15070 | |
| pyridine-2-carboxamide |
| C6H6N2O | |
| MFCD00023483 | |
| picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59 | |
| CHEBI:8200 | |
| NC(=O)C1=CC=CC=N1 |
Specifications
| 1452-77-3 | |
| 97.5% min. (HPLC) | |
| MFCD00023483 | |
| picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59 | |
| NC(=O)C1=CC=CC=N1 | |
| 122.13 | |
| CHEBI:8200 | |
| 98% |
| Authentic | |
| C6H6N2O | |
| 10g | |
| IBBMAWULFFBRKK-UHFFFAOYSA-N | |
| pyridine-2-carboxamide | |
| 15070 | |
| 122.13 | |
| Picolinamide |
Safety and Handling
EINECSNumber : 215-921-5
RUO â Research Use Only