missing translation for 'onlineSavingsMsg'
Learn More

Picolinamide, 98%

Catalog No. AC443360100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443360100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443360100 Supplier Thermo Scientific Chemicals Supplier No. 443360100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1452-77-3 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1452-77-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00023483
InChI Key IBBMAWULFFBRKK-UHFFFAOYSA-N
Synonym picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59
PubChem CID 15070
ChEBI CHEBI:8200
IUPAC Name pyridine-2-carboxamide
SMILES NC(=O)C1=CC=CC=N1

Specifications

Melting Point 106.0°C to 109.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material Picolinamide
EINECSNumber 215-921-5

RUO – Research Use Only