missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picloram 98.0+%, TCI America™
$167.76 - $167.76
Chemical Identifiers
| CAS | 2-1-1918 |
|---|---|
| Molecular Formula | C6H3Cl3N2O2 |
| Molecular Weight (g/mol) | 241.45 |
| MDL Number | MFCD00012101 |
| InChI Key | NQQVFXUMIDALNH-UHFFFAOYSA-N |
| Synonym | picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k |
| PubChem CID | 15965 |
| ChEBI | CHEBI:34922 |
| IUPAC Name | 4-amino-3,5,6-trichloropyridine-2-carboxylic acid |
| SMILES | C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N |
Chemical Identifiers
| 2-1-1918 | |
| 241.45 | |
| NQQVFXUMIDALNH-UHFFFAOYSA-N | |
| 15965 | |
| 4-amino-3,5,6-trichloropyridine-2-carboxylic acid |
| C6H3Cl3N2O2 | |
| MFCD00012101 | |
| picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k | |
| CHEBI:34922 | |
| C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N |
Specifications
| 2-1-1918 | |
| White-Yellow | |
| MFCD00012101 | |
| 3077 | |
| NQQVFXUMIDALNH-UHFFFAOYSA-N | |
| 4-amino-3,5,6-trichloropyridine-2-carboxylic acid | |
| 15965 | |
| 241.45 | |
| Crystalline Powder |
| 219°C | |
| C6H3Cl3N2O2 | |
| 5 g | |
| picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k | |
| C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N | |
| 241.45 | |
| CHEBI:34922 | |
| ≥98.0% (HPLC) | |
| Picloram |
Safety and Handling
RTECSNumber : TJ7525000
TSCA : Yes