missing translation for 'onlineSavingsMsg'
Learn More

Piceid 95.0+%, TCI America™

Catalog No. p-7135303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18781G 1 g
P18785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18781G Supplier TCI America Supplier No. P18781G

Chemical Identifiers

CAS 27208-80-6
Molecular Formula C20H22O8
Molecular Weight (g/mol) 390.388
MDL Number MFCD00210592
InChI Key HSTZMXCBWJGKHG-CUYWLFDKSA-N
PubChem CID 5281718
ChEBI CHEBI:8198
IUPAC Name (2S,3R,4S,5S,6R)-2-[3-hydroxy-5-[(E)-2-(4-hydroxyphenyl)ethenyl]phenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1C=CC2=CC(=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O)O)O

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 1 g
Synonym Resveratrol 3-Glucoside, 4′C,5-Dihydroxystilben-3-yl beta-D-Glucopyranoside, Polydatin
Formula Weight 390.39
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Piceid
TSCA No
Recommended Storage Refrigerator