Learn More
Waters Limited PIC B5 Low UV Reagent DFS Item
Supplier:  Waters Limited WAT084198
Paired -Ion Chromatography (PIC Reagents) provide an easy solution for analyzing samples containing ionic species. These reagents allow ionic compounds to be separated by reverse-phase chromatography, saving time by allowing simultaneous assay of acids, bases, amphoteric and neutral compounds. PIC B5 Low UV is a 5 vial/pkg of pentane sulfonic acid in a workable UV range in mobile phase 200+ nm.
Catalog No. 50785911
Only null left
Explore available promotions