missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phytohemagglutinin, MP Biomedicals™
$128.38 - $128.38
Specifications
| Quantity | 1 mg |
|---|---|
| Physical Form | Powder |
| Chemical Name or Material | Phytohemagglutinin |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15188401
|
MP Biomedicals Inc
0215188401 |
1 mg |
Each for $128.38
|
|
|||||
Specifications
| 1 mg | |
| Powder | |
| Phytohemagglutinin |