missing translation for 'onlineSavingsMsg'
Learn More

Phytantriol (mixture of isomers) 95.0+%, TCI America™

Catalog No. P16745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16745G Supplier TCI America Supplier No. P16745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74563-64-7
Molecular Formula C20H42O3
Molecular Weight (g/mol) 330.55
MDL Number MFCD00129085
InChI Key CGIHFIDULQUVJG-UHFFFAOYNA-N
Synonym 3,7,11,15-Tetramethylhexadecane-1,2,3-triol, 1,2,3-Trihydroxy-3,7,11,15-tetramethylhexadecane
PubChem CID 3018525
ChEBI CHEBI:47770
IUPAC Name 3,7,11,15-tetramethylhexadecane-1,2,3-triol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C(O)CO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 330.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Phytantriol (mixture of isomers)
EINECSNumber (2)-2391
TSCA No