Learn More
Kent Scientific PhysioSuite™ with RightTemp™ Temperature Monitor and Homeothermic Controller
Catalog No. 13005100
Encompass
Change view
Click to view available options
Type:
PhysioSuite™ Monitor
Catalog No. Type
13005100 PhysioSuite™ Monitor
1 options
Catalog No. 13005100 Supplier Kent Scientific Supplier No. PS02
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Homeothermic controller, far infrared warming pad (8 x 10 in.), temperature sensors (mouse and rat sizes), 10 disposable sleeve protectors
Accurately monitors and controls animal temperature.
  • Warms animal with far Infrared heat
  • Safety alarms respond to changes in temperature
  • Modular system integrates physiological monitoring, ventilation, end-tidal CO2 monitoring modules
Specifications
For Use With (Application) Monitors, controls animal temperature
Type PhysioSuite™ Monitor
Includes RightTemp temperature monitor, homeothermic controller