Learn More
Kent Scientific PhysioSuite™ Monitor with CapnoScan™ End-Tidal CO2 Monitor
Catalog No. 13005114
Encompass
Change view
Click to view available options
For Use With (Equipment):
CapnoScan End-Tidal CO2 Monitor
Catalog No. For Use With (Equipment)
13005114 CapnoScan End-Tidal CO2 Monitor
1 options
Catalog No. 13005114 Supplier Kent Scientific Supplier No. PS05
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
RightTemp module
Allows to monitor expired CO2 in intubated rodents.
  • Highly sensitive infrared spectroscopy CO2 sensor
  • RightTemp module included
  • For mice and rats
Specifications
For Use With (Application) Monitor expired CO2 in intubated rodents
Type PhysioSuite™ Monitor
For Use With (Equipment) CapnoScan End-Tidal CO2 Monitor
Includes RightTemp module