missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Physcion, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002022
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002022 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002022 Supplier Merck Emd Millipore Supplier No. PHL8975720MG
Only null left

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 521-61-9
Product Line phyproof
Linear Formula C16H12O5
MDL Number MFCD00017374
Synonym 1,8-Dihydroxy-3-methoxy-6-methylanthraquinone; 6-O-Methylemodin; 6-Methoxychrysophanic acid; Emodin-3-methyl ether; Parietin; Rheochrysidin
Type Reference Substance
Quantity 20 mg
Formula Weight 284.26
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG