missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Isopimpinellin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 482-27-9 |
| Product Line | phyproof |
| Linear Formula | C13H10O5 |
| MDL Number | MFCD00017407 |
| Synonym | 4,9-Dimethoxy-7 H-furo[3,2-g][1 ]benzopyran-7-one; 4,9-Dimethoxyfuro[3,2-g]chromen-7-one; 5,8-Dimethoxy-6,7-furanocoumarin; 5,8-Dimethoxypsoralen; Isoimpinellin; NSC 217988; NSC 401288 |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 246.22 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG