missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Guaiaverin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 22255-13-6 |
| Product Line | phyproof |
| Linear Formula | C20H18O11 |
| Synonym | Quercetin 3-O-alpha-L-arabinopyranoside; 3′,4′,5,7-Tetrahydroxyflavone 3-O-alpha-L-arabinoside; 3-(alpha-L-Arabinopyranosyloxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4 H-1-benzopyran-4-one; Feniculin; Foeniculin; Guaijaverin; Quercetin 3-O-alpha-L-arabinoside |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 434.35 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG