missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Ginsenoside Rg3, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 14197-60-5 |
| Product Line | phyproof |
| Linear Formula | C42H72O13 |
| MDL Number | MFCD06410950 |
| Synonym | (3 beta,12 beta)-12,20-Dihydroxydammar-24-en-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside; 20-Ginsenoside-Rg3; Rg3 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 785.01 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG