missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Formosanin C, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 50773-42-7 |
| Product Line | phyproof |
| Linear Formula | C51H82O20 |
| Synonym | Asperin; Parissaponin; Parissaponin P; Saponin P-d; Saponin Pb |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 1015.18 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG