missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Diosmetin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 520-34-3 |
| Product Line | phyproof |
| Linear Formula | C16H12O6 |
| MDL Number | MFCD00017425 |
| Synonym | 3′,5,7-Trihydroxy-4′-methoxyflavone; 4′-Methylluteolin; 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one; Luteolin 4′-methyl ether |
| Type | Reference Substance |
| Quantity | 25 mg |
| Formula Weight | 300.26 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG