missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Corynoline, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 18797-79-0 |
| Product Line | phyproof |
| Linear Formula | C21H21 NO5 |
| MDL Number | MFCD11042250 |
| Synonym | (+)-Corynoline; 13-Methylchelidonine |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 367.40 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG