missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Alisol B 23-acetate, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 26575-95-1 |
| Product Line | phyproof |
| Linear Formula | C32H50O5 |
| MDL Number | MFCD00210473 |
| Synonym | Alisol B acetate; 23-Acetylalismol B; 23-O-Acetylalisol B; Alisol B 23-acetate |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 514.74 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG