missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ 10-Gingerol, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 23513-15-7 |
| Product Line | phyproof |
| Flash Point | 166.3°C - closed cup |
| Linear Formula | C21H34O4 |
| MDL Number | MFCD01682694 |
| Synonym | [10 ]-Gingerol; - 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone; 10-Gingerol |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 350.49 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG