missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose, 4,000-6,500 cP, NF, 53.4-850%, Spectrum™ Chemical

Catalog No. 18603339
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603339 12 kg Poly Bottle
18603336 125 g Poly Bottle
18603337 500 g Poly Bottle
18603338 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603339 Supplier Spectrum Chemical Mfg Cor Supplier No. HY12112KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HY121, 9004-64-2

Spectrum™ Chemical Hydroxypropyl Cellulose, 4,000-6,500 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
SMILES CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*

Specifications

CAS Max % 100%
Ignition Residue 0.8%
Loss on Drying 5%
Packaging Poly Bottle
Quantity 12 kg
Molecular Weight (g/mol) 850
Percent Purity 53.4 to 80.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Hydroxypropyl Cellulose, 4,000-6,500 cP