missing translation for 'onlineSavingsMsg'
Learn More

Phthaloyl Chloride 98.0+%, TCI America™

Catalog No. p-7134591
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P040525G 25 g
P0405100G 100 g
P0405500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P040525G Supplier TCI America Supplier No. P040525G
Only null left

Chemical Identifiers

CAS 88-95-9
Molecular Formula C8H4Cl2O2
Molecular Weight (g/mol) 203.018
MDL Number MFCD00000666
InChI Key FYXKZNLBZKRYSS-UHFFFAOYSA-N
Synonym phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
PubChem CID 6955
IUPAC Name benzene-1,2-dicarbonyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl

Specifications

Color Yellow
Boiling Point 276°C
Quantity 25 g
UN Number 3265
Formula Weight 203.02
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Phthaloyl Chloride
EINECSNumber (3)-1378&(3)-2973
TSCA Yes