missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalonitrile 99.0+%, TCI America™
Supplier: TCI America P0404500G
Specifications
| Phthalonitrile | |
| 140°C | |
| C8H4N2 | |
| 500 g | |
| phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile | |
| N#CC1=CC=CC=C1C#N | |
| 128.13 | |
| 128.13 | |
| Crystalline Powder |
| 91-15-6 | |
| White | |
| MFCD00001771 | |
| 3439 | |
| XQZYPMVTSDWCCE-UHFFFAOYSA-N | |
| benzene-1,2-dicarbonitrile | |
| 7042 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 91-15-6 | |
| 128.13 | |
| XQZYPMVTSDWCCE-UHFFFAOYSA-N | |
| 7042 | |
| N#CC1=CC=CC=C1C#N |
| C8H4N2 | |
| MFCD00001771 | |
| phthalonitrile, 1,2-dicyanobenzene, o-phthalodinitrile, phthalodinitrile, 1,2-benzenedicarbonitrile, o-dicyanobenzene, 1,2-benzodinitrile, o-pdn, o-cyanobenzonitrile, o-benzenedicarbonitrile | |
| benzene-1,2-dicarbonitrile |
Safety and Handling
EINECSNumber : (3)-1799
RTECSNumber : TI8575000
TSCA : Yes